Learn More
Cole-Parmer Cole-Parmer Stainless steel double-ended spoon set, Set of 3
Catalog No. NC9866820 Shop All Cole-Parmer Products
Encompass
Double-ended spoons feature a solid rounded shaft that is comfortable to hold for long periods. Set contains three spoons: a 5-7/8" (14.9 cm) long spoon with 0.5 mL and 2.0 mL bowls, an 8-1/4" (21.0 cm) long spoon with 1.6 mL and 5.0 mL bowls, and an 11-7/8"(30.2 cm) long spoon with 2.0 mL and 8.0 mL bowls.
Catalog No. NC9866820 Supplier Cole-Parmer Supplier No. 0626100
Double-ended spoons feature a solid rounded shaft that is comfortable to hold for long periods. Set contains three spoons: a 5-7/8" (14.9 cm) long spoon with 0.5 mL and 2.0 mL bowls, an 8-1/4" (21.0 cm) long spoon with 1.6 mL and 5.0 mL bowls, and an 11-7/8"(30.2 cm) long spoon with 2.0 mL and 8.0 mL bowls.