Learn More
Sigma Aldrich Fine Chemicals Biosciences Double-ended needle gauge
Catalog No. 502900219
Encompass
304 SS. One noncoring tip one flat-cut end.
Catalog No. 50-290-0219 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1163946EA
304 SS. One noncoring tip one flat-cut end.