Learn More
Sklar Instruments Double Ended Probe, Sklar, 8 in., 1mm, Straight, Rounded, Stainless Steel. EA/1.
SDP
Catalog No. 501438124 Shop All Sklar Instruments Products
Encompass_Preferred
Double Ended Probe, Sklar, 8 in., 1mm, Straight, Rounded, Stainless Steel. Sklar's Double Ended Probe is an instrument used to assist in retrieving information such as depth or size. The probe is used to determine weather foreign bodies or other issues exist. The probe is available in a variety of lengths and tip sizes.
This item is not returnable. View return policy
Catalog No. 50-143-8124 Supplier Sklar Instruments Supplier No. 301512
Double Ended Probe, Sklar, 8 in., 1mm, Straight, Rounded, Stainless Steel. Sklar's Double Ended Probe is an instrument used to assist in retrieving information such as depth or size. The probe is used to determine weather foreign bodies or other issues exist. The probe is available in a variety of lengths and tip sizes.