Learn More
Med Vet International Double Ended Subgingival Curette, Stainless Steel, 6”, J0042S
Catalog No. 5000585168
Encompass_Preferred
Gracey style. Subgingival curette for important removal of necrotic debris below the gumline. Often overlooked but necessary for proper dental care. England.
Catalog No. 50-005-85168 Supplier Med Vet International Supplier No. JOR42S
Gracey style. Subgingival curette for important removal of necrotic debris below the gumline. Often overlooked but necessary for proper dental care. England.