Learn More
Cole-Parmer Sellstrom 31140 Laboratory faceshield, double crown w/acetate shield
Catalog No. NC9345038 Shop All Cole-Parmer Products
Encompass_Preferred
White ABS plastic crown includes ratchet headgear and deluxe perspiration band to maintain a cool, comfortable fit. Lightweight clear acetate window (included) is fog-free to allow maximum visibility. Ideal for laboratory use. Select single-crown faceshield for protection from splash and particles. Select double-crown faceshield with chin protector for added impact protection.
This item is not returnable. View return policy
Catalog No. NC9345038 Supplier Cole-Parmer Supplier No. 8647822
White ABS plastic crown includes ratchet headgear and deluxe perspiration band to maintain a cool, comfortable fit. Lightweight clear acetate window (included) is fog-free to allow maximum visibility. Ideal for laboratory use. Select single-crown faceshield for protection from splash and particles. Select double-crown faceshield with chin protector for added impact protection.