Learn More
Justrite Double Gas-Cylinder Hand Trucks
Catalog No. 19085761 Shop All Justrite Safety Group Products
Change view
Click to view available options
Holds:
2 Cylinders
Catalog No. Holds
19-085-761 2 Cylinders
1 options
Catalog No. 19-085-761 Supplier Justrite Supplier No. 35032
Add to Cart
Add to Cart
Specially designed to load and transport heavy gas-cylinders and minimize accidents or injuries.
  • Constructed of 1 in. (25 mm) and 1.25 in. (32 mm) diameter steel tubing with continuous welding
  • 11-gauge (3 mm) steel floor plate with safety fins prevents warping or breaking
  • The unique zero-lift height floor plate lays flat, eliminates the need to lift cylinder during loading and unloading
  • Corrosion-resistant powder coat finish for long life
  • Heavy-duty 1.5 in. (38 mm) wide poly straps, lab tested to 1138 lb. (516 kg), with steel cinch buckles to eliminate potential cylinder rolling
  • All main wheels have dual ball bearings for easier rolling
Specifications
Type Gas Cylinder Cart
Holds 2 Cylinders
Load Bearing Capacity (English) 1000 lb.
Load Bearing Capacity (Metric) 454 kg
Material Steel