Learn More
Jorgensen Labs Double Guarded Culture Swab, 25/pk
SDP
Catalog No. NC0444327
Encompass_Preferred
This polystyrene cotton swab is protected by two outer plastic sheaths and a cap. The outer sheath and cap are penetrated after the swab passes the cervix into the uterus. The swab is scored so its length can be reduced and then covered with the two included caps for storage or transport. 33″L. 25/pk.
Catalog No. NC0444327 Supplier Jorgensen Labs Supplier No. J0273
This polystyrene cotton swab is protected by two outer plastic sheaths and a cap. The outer sheath and cap are penetrated after the swab passes the cervix into the uterus. The swab is scored so its length can be reduced and then covered with the two included caps for storage or transport. 33″L. 25/pk.