Learn More
Brady™ Double Hex Prolock
Catalog No. 19103918 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Red
Catalog No. Color
19-103-918 Red
1 options
Catalog No. 19-103-918 Supplier Brady™ Supplier No. 122247
  • $31.50
    $24.65 / Each of 1
    Save $6.85
    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Effectively lock out a variety of isolation points. The Brady™ Double Hex Prolock will quickly become a staple lockout device in any facility. It tackles anything from T valve handles to electrical disconnects.
  • Designed to effectively lock out a variety of isolation points, including gate valves, T valve handles, electrical disconneccts, and more
  • Economical and commact devices are highly visible and will quickly become a staple lockout device in any facility
Specifications
Product Type Pull-tight Cable Lockout Device
Color Red
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.