Learn More
Euthanex Integrated Imaging Double Breather Block
Catalog No. 501540153
Encompass_Preferred
Integrated Imaging Double Breather Block can be permanentely integrated into a wide range of imaging systems. The low profile, all plastic construction breathers include threaded mounting holes for simple integration. Designed for use with animals weighing up to 500 grams. Includes all tubing and connectors.
Catalog No. 50-154-0153 Supplier Euthanex Supplier No. EZ109012
Integrated Imaging Double Breather Block can be permanentely integrated into a wide range of imaging systems. The low profile, all plastic construction breathers include threaded mounting holes for simple integration. Designed for use with animals weighing up to 500 grams. Includes all tubing and connectors.