Learn More
Braintree Scientific In Vivo Perfusion System for mice (double) 140ml - IV-4140
SDP
Catalog No. NC9069235
Encompass
AutoMate In Vivo Manual Gravity Perfusion System. Take the guesswork out of your animal fixation system. Providing an option for scientists who would rather do science than design a system and source parts. Let AutoMate take the guesswork out of your new animal fixation system. Steady gravity-fed delivery of buffer and fixative, results in complete perfusion. Easy to set up, use and clean. Complete system with everything down to the needles. Fits in standard hoods at a fraction of the cost of a peristaltic pump and more reliable. One or two users can perfuse two mice simultaneously with the Double In Vivo system. Four reservoirs and two stopcocks are mounted for two independent wash buffer and fixative deliveries on the same ringstands. 2-way manual stopcock makes for easy one-handed operation to switch solutions. **Sold for research purposes only and not for human use**
Catalog No. NC9069235 Supplier Braintree Scientific Supplier No. IV4140
 Restricted Item
Add to Cart
Add to Cart
AutoMate In Vivo Manual Gravity Perfusion System. Take the guesswork out of your animal fixation system. Providing an option for scientists who would rather do science than design a system and source parts. Let AutoMate take the guesswork out of your new animal fixation system. Steady gravity-fed delivery of buffer and fixative, results in complete perfusion. Easy to set up, use and clean. Complete system with everything down to the needles. Fits in standard hoods at a fraction of the cost of a peristaltic pump and more reliable. One or two users can perfuse two mice simultaneously with the Double In Vivo system. Four reservoirs and two stopcocks are mounted for two independent wash buffer and fixative deliveries on the same ringstands. 2-way manual stopcock makes for easy one-handed operation to switch solutions. **Sold for research purposes only and not for human use**