Learn More
Cole-Parmer Oakton General-Purpose ORP Probe, Sealed, DJ, Epoxy, 3 ft Cable, BNC
Catalog No. NC0915546 Shop All Cole-Parmer Products
Encompass_Preferred
Compatible with all Oakton brand pH meters. Ideal for ORP dirty sample applications. Impact resistant and double junction type.
Catalog No. NC0915546 Supplier Cole-Parmer Supplier No. 3580515
Compatible with all Oakton brand pH meters. Ideal for ORP dirty sample applications. Impact resistant and double junction type.