Learn More
Cole-Parmer Oakton General-Purpose pH Probe, Sealed, DJ, Epoxy, 3 ft Cable, BNC
Catalog No. NC9807201 Shop All Cole-Parmer Products
Encompass_Preferred
Oakton combination electrodes are available in a variety of styles to meet the needs of your application. Epoxy electrodes are impact resistant and provide excellent durability for field applications. Glass electrodes withstand aggressive chemicals. Choose junction and tip style based on the needs of your application. All electrodes feature Ag/AgCl reference cells.
Catalog No. NC9807201 Supplier Cole-Parmer Supplier No. 3580501
Oakton combination electrodes are available in a variety of styles to meet the needs of your application. Epoxy electrodes are impact resistant and provide excellent durability for field applications. Glass electrodes withstand aggressive chemicals. Choose junction and tip style based on the needs of your application. All electrodes feature Ag/AgCl reference cells.