Learn More
Vector Laboratories VectaFluor Horse Anti-Rabbit DK-8828
SDP
Catalog No. 502045644
Encompass
VectaFluor Horse Anti-Rabbit IgG, DyLight 594 Antibody Kit, R.T.U., 1 Kit. Ready-to-use (R.T.U.) format offers maximum convenience for fluorescence staining of cells and tissues. This affinity-purified, ultrapure, high-affinity antibody has been thoroughly adsorbed against serum and immunoglobulins from potentially interfering species. Unless otherwise specified, antibodies recognize both heavy and light chains (H+L). Optimally labeled with DyLight 594 to provide the brightest label for fluorescence microscopy. Excitation: 592 nm; Emission: 617 nm; Color: Red. Kit Contents: 15 ml pre-diluted, stabilized secondary antibody solution 15; ml 2.5% normal horse serum for blocking.
Catalog No. 50-204-5644 Supplier Vector Laboratories Supplier No. DK8828
VectaFluor Horse Anti-Rabbit IgG, DyLight 594 Antibody Kit, R.T.U., 1 Kit. Ready-to-use (R.T.U.) format offers maximum convenience for fluorescence staining of cells and tissues. This affinity-purified, ultrapure, high-affinity antibody has been thoroughly adsorbed against serum and immunoglobulins from potentially interfering species. Unless otherwise specified, antibodies recognize both heavy and light chains (H+L). Optimally labeled with DyLight 594 to provide the brightest label for fluorescence microscopy. Excitation: 592 nm; Emission: 617 nm; Color: Red. Kit Contents: 15 ml pre-diluted, stabilized secondary antibody solution 15; ml 2.5% normal horse serum for blocking.