Learn More
Braintree Scientific Double lumen Intracranial Catheter11.7cm 32g accepts 32g ndl
SDP
Catalog No. 501954585
Encompass
Double lumen intracranial catheter system contains an 11.7cm polyurethane catheter, a bonded connection assembly flared to accept needle. A 0.003" diameter Teflon coated stainless steel stylet to facilitate placement. Individually packaged. Supplied non-sterile. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4585 Supplier Braintree Scientific Supplier No. CRDL32
Double lumen intracranial catheter system contains an 11.7cm polyurethane catheter, a bonded connection assembly flared to accept needle. A 0.003" diameter Teflon coated stainless steel stylet to facilitate placement. Individually packaged. Supplied non-sterile. **Sold for research purposes only and not for human use**