Learn More
Synthware™ Double Manifold with High Vacuum Valve and Spherical Joint
GREENER_CHOICESDP
Catalog No. 31501803 Shop All Kemtech Products
Change view
Click to view available options
Material:
Borosilicate Glass
Catalog No. Material
31-501-803 Borosilicate Glass
1 options
Catalog No. 31-501-803 Supplier Synthware™ Supplier No. M360004
Add to Cart
Add to Cart
Designed to incorporate a 0 to 8 mm high vacuum valve as a vacuum shut off. The front tube is for inert gases (17 mm OD), with a 10 mm hose connection. Rear tube is for vacuum (22 mm OD) with two 35/20 socket joints.
Specifications
Description Front Right; Rear Left and Right Spherical Connection
Type Double Manifold
For Use With (Application) Organic Synthesis
Includes 4 ports
Material Borosilicate Glass