Learn More
Synthware™ Double Manifold with Hollow Glass Stopcocks
GREENER_CHOICESDP
Catalog No. 31501741 Shop All Kemtech Products
Change view
Click to view available options
Port Size (Metric):
4 ports
Catalog No. Port Size (Metric)
31-501-741 4 ports
1 options
Catalog No. 31-501-741 Supplier Synthware™ Supplier No. M471004
Add to Cart
Add to Cart
General purpose double bank manifold for use with vacuum and inert gas. Supplied with 0 to 4 mm high vacuum valves for grease-free operation. Both banks are made from 22 mm medium wall tubing and have all banks open. Ports are spaced 100 mm center to center and have an O.D. of 10 mm.
Front Left, Rear Left and Right Hose Connection
Specifications
Type Vacuum/Inert Gas Double Manifold
For Use With (Application) Organic Synthesis
Material Borosilicate Glass
Port Size (Metric) 4 ports
Width (Metric) 450 mm