Learn More
MSC Kwazar Spray Bottle, Mist Dispensing, 1 L, Black, 1 ea
Catalog No. 502867808
Encompass
This plastic spray bottle features a trigger-activated mist dispensing system and a 1 L container capacity, designed for efficient application in various settings.
  • Delivers 1.22 mL per squeeze with continuous output to reduce effort
  • Ergonomic design prevents hand fatigue during extended use
  • Durable spray head resists acids and disinfectants
  • Adjustable nozzle provides spray from super-fine mist to powerful jet stream
  • Suitable for dairy, equine, and general agricultural applications
  • Equipped with inline filters and a balanced shape for stability
  • Screw closure size is 24/415
  • Fill opening diameter is 20 mm
  • Dip tube length is 9-1/2 in
Catalog No. 50-286-7808 Supplier MSC Supplier No. 74253147
This plastic spray bottle features a trigger-activated mist dispensing system and a 1 L container capacity, designed for efficient application in various settings.
  • Delivers 1.22 mL per squeeze with continuous output to reduce effort
  • Ergonomic design prevents hand fatigue during extended use
  • Durable spray head resists acids and disinfectants
  • Adjustable nozzle provides spray from super-fine mist to powerful jet stream
  • Suitable for dairy, equine, and general agricultural applications
  • Equipped with inline filters and a balanced shape for stability
  • Screw closure size is 24/415
  • Fill opening diameter is 20 mm
  • Dip tube length is 9-1/2 in