Learn More
Chemglass Life Sciences Stopcock, Double Oblique Bore, PTFE Plug, 4mm Bore, 14.4/44 Plug Size, 10mm OD Sidearms
SDP
Catalog No. CG44604
Encompass_Preferred
Stopcock, Double Oblique Bore, PTFE Plug, 4mm Bore, 14.4/44 Plug Size, 10mm OD Sidearms. A 1:5 taper PTFE stopcock with two parallel diagonal bores.
Catalog No. CG44604 Supplier Chemglass Life Sciences Supplier No. CG44604
Stopcock, Double Oblique Bore, PTFE Plug, 4mm Bore, 14.4/44 Plug Size, 10mm OD Sidearms. A 1:5 taper PTFE stopcock with two parallel diagonal bores.