Learn More
Electron Microscopy Sciences Gilder Double Oyster Grid 50/100, Nickel
SDP
Catalog No. 5025620
Encompass_Preferred
Double grids are used mostly in metallurgical applications for supporting thin metal foils. These grids have a curved securing tab which folds to the curvature of the 'sandwiched' grid. 3.05 mm diameter, 0.7 mil (18um) thickness.
Material: Nickel
Pitch: 500/250 µm
Hole: 430/195 µm
Bar: 70/55 µm
Catalog No. 50-256-20 Supplier Electron Microscopy Sciences Supplier No. GD5010NI
Double grids are used mostly in metallurgical applications for supporting thin metal foils. These grids have a curved securing tab which folds to the curvature of the 'sandwiched' grid. 3.05 mm diameter, 0.7 mil (18um) thickness.
Material: Nickel
Pitch: 500/250 µm
Hole: 430/195 µm
Bar: 70/55 µm