Learn More
Research Products International Corp Orbital Benchtop Shaker with Double Stacked Platform, 10 1/4 x 9 3/4 x 4 3/4 H Inches
SDP
Supplier:  Research Products International Corp 248910
Encompass
Versatile orbital shaker features interchangeable platforms to accommodate tubes, dishes, trays or flasks. Shaker contains a powerful brushless motor with a counter balanced mechanism for smooth, quiet and dependable service. Adjustable speed control can be set from 20-300 rpm.
12 month warranty.
Catalog No. 50-212-654