Learn More
Mopec Double-pouch Specimen Bag
SDP
Catalog No. 22446148 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
6 x 10 in.
Includes:
Absorbing Sheet
No Absorbing Sheet
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Includes
22-446-148 6 x 10 in. Absorbing Sheet
22-446-109 6 x 10 in. No Absorbing Sheet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-446-148 Supplier Mopec Supplier No. FS084
Add to Cart
Add to Cart
Designed to securely contain leaks from urine bottles, featuring a Tamper-evident adhesive seal that cannot be reopened without damage.
  • Contains liquid in the event of a urine bottle leak, with absorbent sheets capable of absorbing the entire contents of a specimen bottle, while keeping the specimen bottles separate from documentation.
  • Features dual compartments to separate specimens from paperwork.
  • Available with optional absorbent sheets that gel on contact with liquid.
  • Absorbent sheets can handle anything from minor spills to full bottle contents.
Specifications
Dimensions (L x W) 6 x 10 in.
Product Type Speciman Bag
Includes Absorbing Sheet
Color Transparent with Black Print
Quantity 200/Pk.