Learn More
Mitegen Double Puck Loading Dewar with Lid, Holds 2 Uni-Pucks or Cryo-EM pucks, Double-Sided, 14"x7"x8", 0.75 lbs, For Cryogenic Puck Handling
SDP
Catalog No. 501187179
Encompass
This Double Puck Loading Dewar with Lid is specifically designed to streamline the handling and organization of cryogenic samples in the laboratory. It features space to securely hold two Uni-Pucks or Cryo-EM pucks, offering double-sided access for efficient sample management. With compact dimensions of 14 inches x 7 inches x 8 inches and a lightweight design weighing only 0.75 lbs, this dewar is highly portable and easy to manipulate during critical sample transfers. The included lid ensures the integrity of the cryogenic environment, providing an essential barrier for maintaining ultra-low temperatures and preventing contamination. Ideal for use in cryo-crystallography and other cryo-EM workflows, this dewar is an indispensable tool for preparing and transporting multiple samples with enhanced safety and organization. It significantly improves workflow efficiency in demanding cryogenic environments. Use in conjunction with M-CEM-INS1 for Cryo-EM Pucks
Catalog No. 50-118-7179 Supplier Mitegen Supplier No. MCP111022
This Double Puck Loading Dewar with Lid is specifically designed to streamline the handling and organization of cryogenic samples in the laboratory. It features space to securely hold two Uni-Pucks or Cryo-EM pucks, offering double-sided access for efficient sample management. With compact dimensions of 14 inches x 7 inches x 8 inches and a lightweight design weighing only 0.75 lbs, this dewar is highly portable and easy to manipulate during critical sample transfers. The included lid ensures the integrity of the cryogenic environment, providing an essential barrier for maintaining ultra-low temperatures and preventing contamination. Ideal for use in cryo-crystallography and other cryo-EM workflows, this dewar is an indispensable tool for preparing and transporting multiple samples with enhanced safety and organization. It significantly improves workflow efficiency in demanding cryogenic environments. Use in conjunction with M-CEM-INS1 for Cryo-EM Pucks