Learn More
Mitegen Double Puck Loading Foam Dewar with Lid and Cryo-EM Puck Holder Insert
SDP
Catalog No. 5000494984
Encompass
Foam double puck loading dewar (lid included) with Cryo-EM Puck Holder Insert for easy grid transfer. Easily transfer grid boxes to and from pucks while immersed in LN2. Insert is form fitted to the double puck loading dewar to securely hold Cryo-EM puck in place. Engraved markings on the insert helps locate and index positions of grid box locations for logging and tracking.
Catalog No. 50-004-94984 Supplier Mitegen Supplier No. MCEMPDIS1
Foam double puck loading dewar (lid included) with Cryo-EM Puck Holder Insert for easy grid transfer. Easily transfer grid boxes to and from pucks while immersed in LN2. Insert is form fitted to the double puck loading dewar to securely hold Cryo-EM puck in place. Engraved markings on the insert helps locate and index positions of grid box locations for logging and tracking.