Learn More
VELP Scientifica™ Double Rod Mini H-Stand Set
SDP
Catalog No. 08418525
Change view
Click to view available options
Dimensions (L x W x H):
300 x 430 mm
Catalog No. Dimensions (L x W x H)
08-418-525 300 x 430 mm
1 options
Catalog No. 08-418-525 Supplier VELP Scientifica™ Supplier No. A00000383
The double rod mini H-stand set for disperser tools is designed for light weight instruments and comes complete. This stand is designed to avoid rotation of attachments and instruments.
DESCRIPTION
  • Designed for light weight instruments and comes complete with all parts
  • Designed to avoid rotation of attachment and instrument through the use of the double rod and bosshead clamp
Specifications
Product Type Stand
Dimensions (L x W x H) 300 x 430 mm
Autoclavable Not Autoclavable