Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Conductive Double Sided Carbon Tape, 20 mm
SDP

Catalog No. 5028585
Encompass_Preferred

Can be used to mount particles 15-20 microns. Isolate unwoved cloth. Adhesive power/25mm: 1000 gf.
Conductive resistivity: 50ohm/sq.Inch
Thickness: 0.16 mm
Pad: 0.07 mm
Adhesive: 0.045 mm x 2

Catalog No. 50-285-85 Supplier Electron Microscopy Sciences Supplier No. 7781720
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Can be used to mount particles 15-20 microns. Isolate unwoved cloth. Adhesive power/25mm: 1000 gf.
Conductive resistivity: 50ohm/sq.Inch
Thickness: 0.16 mm
Pad: 0.07 mm
Adhesive: 0.045 mm x 2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.