Description
Abrasion resistant polyester knit clean room fabric laminated to absorbent polyurethane foam. Laser cut and cleanroom laundered to minimize fiber and particle release. 10 mm thickness provides cushioned surface for optimum conformance to uneven floors, walls and ceilings. High absorbency capacity.
- Abrasion resistant
- Laser cut
- High absorbency
Specifications
Specifications
| Product Type | Double-sided Mop Cover |
| Sterility | Non-sterile |
| Color-coded | No |
| Material | Foam |