Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC AccuformNMC Floor Sign, Double-Sided, Polyethylene, 12 x 20 in

Catalog No. 502836467
Encompass

This double-sided floor sign provides clear temporary hazard warnings for industrial and commercial environments. Constructed from durable, waterproof polyethylene, it features two distinct safety messages for optimal visibility. Its lightweight and foldable A-frame design ensures easy deployment, transport, and storage.

  • Double-sided for clear visibility from multiple directions.
  • Constructed from polyethylene for durability and weather resistance.
  • Front message: "CAUTION KEEP CLEAR MACHINE UNDER TEST".
  • Back message: "CAUTION KEEP CLEAR HAZARDOUS AREA".
  • Measures 12 inches wide by 20 inches high.
  • Economical and lightweight design.
  • Waterproof and easily foldable for storage.
  • A-frame shape for stable floor mounting.

Catalog No. 50-283-6467 Supplier MSC Supplier No. 74725862
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This double-sided floor sign provides clear temporary hazard warnings for industrial and commercial environments. Constructed from durable, waterproof polyethylene, it features two distinct safety messages for optimal visibility. Its lightweight and foldable A-frame design ensures easy deployment, transport, and storage.

  • Double-sided for clear visibility from multiple directions.
  • Constructed from polyethylene for durability and weather resistance.
  • Front message: "CAUTION KEEP CLEAR MACHINE UNDER TEST".
  • Back message: "CAUTION KEEP CLEAR HAZARDOUS AREA".
  • Measures 12 inches wide by 20 inches high.
  • Economical and lightweight design.
  • Waterproof and easily foldable for storage.
  • A-frame shape for stable floor mounting.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.