Learn More
Grainger Double Sided Foam Tape 1 L 1 W PK256
Catalog No. 502766744
Encompass_Preferred
The 3M Double Sided Foam Tape 16U614 is a high performance acrylic adhesive tape with a polyethylene foam carrier. It has a thickness of 1/16 inch and is available in rolls that are 36 yards long and 1 inch wide. The tape has excellent adhesion to a variety of surfaces including wood, glass, plastic, and metal. It bonds on contact and offers high shear strength while conforming to uneven surfaces. This versatile tape is ideal for mounting signs, nameplates, moldings, and trims.
Catalog No. 50-276-6744 Supplier Grainger Supplier No. 16U614
The 3M Double Sided Foam Tape 16U614 is a high performance acrylic adhesive tape with a polyethylene foam carrier. It has a thickness of 1/16 inch and is available in rolls that are 36 yards long and 1 inch wide. The tape has excellent adhesion to a variety of surfaces including wood, glass, plastic, and metal. It bonds on contact and offers high shear strength while conforming to uneven surfaces. This versatile tape is ideal for mounting signs, nameplates, moldings, and trims.