Learn More
Grainger Double Sided Foam Tape 72 yd L 1 W
Catalog No. 502752817
Encompass_Preferred
The 3M Double Sided Foam Tape is a black, double-sided acrylic foam tape offering high strength bonding for various materials. It is 1/16 inch thick, 36 yards long, and 1 inch wide. The acrylic adhesive ensures excellent adhesion to surfaces like wood, glass, plastic, and metal. It bonds on contact for easy bonding, mounting, and fastening. Suitable for indoor and outdoor use, it withstands UV light and has a service temperature range of -40°F to 200°F. Key features include high strength adhesive, quick stick at room temperature, and resistance to UV light, solvents, and moisture.
Catalog No. 50-275-2817 Supplier Grainger Supplier No. 15D143
The 3M Double Sided Foam Tape is a black, double-sided acrylic foam tape offering high strength bonding for various materials. It is 1/16 inch thick, 36 yards long, and 1 inch wide. The acrylic adhesive ensures excellent adhesion to surfaces like wood, glass, plastic, and metal. It bonds on contact for easy bonding, mounting, and fastening. Suitable for indoor and outdoor use, it withstands UV light and has a service temperature range of -40°F to 200°F. Key features include high strength adhesive, quick stick at room temperature, and resistance to UV light, solvents, and moisture.