Learn More
Grainger Acrylic Foam Double Sided Foam Tape, Acrylic Adhesive, 40 mil Thick, 3/4 in X 5 1/2 yd, Clear
Catalog No. NC1258961
Encompass_Preferred
SilverTape 5310 is designed for bonding materials with medium and high surface energy where high transparency is desired, such as glass and transparent plastics. The tape is capable of absorbing the differing thermal expansion rates of two materials, and has high impact resistance even at temperatures below 32°F (0°C).
Catalog No. NC1258961 Supplier Grainger Supplier No. 36LN06
SilverTape 5310 is designed for bonding materials with medium and high surface energy where high transparency is desired, such as glass and transparent plastics. The tape is capable of absorbing the differing thermal expansion rates of two materials, and has high impact resistance even at temperatures below 32°F (0°C).