Learn More
OHAUS Double Sided Tape Roll
Catalog No. 01804374 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Shaker Attachment
Catalog No. Product Type
01-804-374 Shaker Attachment
1 options
Catalog No. 01-804-374 Supplier OHAUS Supplier No. 83041437
Double Sided Adhesive Tape is an alternative to flask clamps or roller bars for securing vessels to shaker platform of Orbital-Genie Shaker.
  • It is ideal for containers such as bottles and dishes that cannot be held with conventional clamps or roller bars.
  • Cut strips of tape as desired and attach to platform.
  • Can only be used with SI-16xx or SI-17xx.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Shaker Attachment