Learn More
Advanced Medical Systems EnviroTest Kit 10/case.
Catalog No. NC0204810
Encompass_Preferred
Promotes rapid growth of common bacteria, yeast, and moldNeutralizes many residual biocides and cleaning agents -Stores at room temperature -Long shelf life without refrigeration -Unique pedestal cap and hinged paddle -Log sheet included -Gummed labels included
Catalog No. NC0204810 Supplier Advanced Medical Systems Supplier No. PMTD100
Promotes rapid growth of common bacteria, yeast, and moldNeutralizes many residual biocides and cleaning agents -Stores at room temperature -Long shelf life without refrigeration -Unique pedestal cap and hinged paddle -Log sheet included -Gummed labels included