Learn More
Grainger 3M VHB Tape, Acrylic Foam, Double-Sided, 25 Mil
Catalog No. 502756517
Encompass_Preferred
This double-sided, general-purpose acrylic tape features a firm foam core designed to replace mechanical fasteners like rivets, welds, and screws. Providing a permanent bond on contact, it allows for faster production by eliminating drilling, grinding, and cleanup steps. The closed-cell acrylic technology ensures a durable, weather-resistant seal for indoor and outdoor applications on a wide range of high surface energy substrates.
  • Provides immediate handling strength with no required curing time
  • Enhances finished goods appearance through seamless, invisible joining
  • Enables use of thinner, lightweight, and dissimilar materials
  • Resists temperatures up to 300 °F (149 °C) for short-term and 200 °F (93 °C) for long-term exposure
  • Offers excellent moisture and solvent resistance for long-term durability
Catalog No. 50-275-6517 Supplier Grainger Supplier No. 15C322
This double-sided, general-purpose acrylic tape features a firm foam core designed to replace mechanical fasteners like rivets, welds, and screws. Providing a permanent bond on contact, it allows for faster production by eliminating drilling, grinding, and cleanup steps. The closed-cell acrylic technology ensures a durable, weather-resistant seal for indoor and outdoor applications on a wide range of high surface energy substrates.
  • Provides immediate handling strength with no required curing time
  • Enhances finished goods appearance through seamless, invisible joining
  • Enables use of thinner, lightweight, and dissimilar materials
  • Resists temperatures up to 300 °F (149 °C) for short-term and 200 °F (93 °C) for long-term exposure
  • Offers excellent moisture and solvent resistance for long-term durability