Learn More
Cell Signaling Technology Double Strand Breaks Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502729737
Encompass
The Double Strand Breaks (DSB) Repair Antibody Sampler Kit provides an economical means to investigate the repair of double-strand DNA breaks within the cell. This kit contains a selection of primary and secondary antibodies curated to facilitate the analysis of DNA damage response pathways, including non-homologous end-joining and homologous recombination, through western blotting.
  • Includes antibodies against key DNA damage response proteins such as ATM, Mre11, Rad50, DNA-PK, and Ku80
  • Features both phospho-specific and total protein targets to study activation states
  • Supplied with sufficient antibody quantity to perform two western blots per target
  • Designed for research applications in DNA damage and senescence
Catalog No. 50-272-9737 Supplier Cell Signaling Technology Supplier No. 9653T
The Double Strand Breaks (DSB) Repair Antibody Sampler Kit provides an economical means to investigate the repair of double-strand DNA breaks within the cell. This kit contains a selection of primary and secondary antibodies curated to facilitate the analysis of DNA damage response pathways, including non-homologous end-joining and homologous recombination, through western blotting.
  • Includes antibodies against key DNA damage response proteins such as ATM, Mre11, Rad50, DNA-PK, and Ku80
  • Features both phospho-specific and total protein targets to study activation states
  • Supplied with sufficient antibody quantity to perform two western blots per target
  • Designed for research applications in DNA damage and senescence