Learn More
Genscript Corporation Double-stranded RNA dsRNA An
Catalog No. 5000058897
Encompass
This product is specific for double-stranded RNA (dsRNA) with the length of the helix greater than or equal to 40 bp The dsRNA-recognition is independent of the sequence and nucleotide composition of the antigen
Catalog No. 50-000-58897 Supplier Genscript Corporation Supplier No. A0218240
This product is specific for double-stranded RNA (dsRNA) with the length of the helix greater than or equal to 40 bp The dsRNA-recognition is independent of the sequence and nucleotide composition of the antigen