Learn More
Ace Glass, Inc. DOUBLE TUBE MANIFOLD DOUBLE TUBE MANIFOLD
SDP
Catalog No. 50975462
Encompass_Preferred
Double tube manifold with (4) 4mm double oblique stopcocks for ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-975-462 Supplier Ace Glass, Inc. Supplier No. 781824
Double tube manifold with (4) 4mm double oblique stopcocks for ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.