Learn More
Thomas Scientific Counter, Double
SDP
Catalog No. NC9234755
Encompass
Used for blood cell and corpuscle counts or wherever double counting is required Two registers each registers to 9999 Large, rectangular pushbuttons and zero reset knob on both sides Pushbuttons for the two independent registers are color-coded, and both sides are reset simultaneously by either knob
Catalog No. NC9234755 Supplier Thomas Scientific Supplier No. 3297J60
Used for blood cell and corpuscle counts or wherever double counting is required Two registers each registers to 9999 Large, rectangular pushbuttons and zero reset knob on both sides Pushbuttons for the two independent registers are color-coded, and both sides are reset simultaneously by either knob