Learn More
Description
- A small diameter inner tube extends down the annular space to ensure flow or coolant from bottom of well, upward to outlet
- Use with RC50 Nylon™ Bushing and a Viton™ O-ring to form a compression type seal with threaded vessels
- Lack of ground joint means dismantling since there is no freezing
- Well can be adjusted to any desired height
- Greaseless connection
Specifications
Specifications
| Capacity (Metric) | 390 mL |
| Material | Borosilicate Glass (3.3) |
| For Use With (Equipment) | V150500 Reactor and Lamp |
| Product Type | Immersion Well |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | No |
| Mouth Style | Joint |
| Sterility | Non-sterile |
| Bottom Shape | Round |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

