Learn More
Thermo Scientific™ Double Zip AnaeroPouch™-Bag
GSA_VA
Catalog No. 23246390 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20/Pk.
Catalog No. Quantity
23-246-390 20/Pk.
1 options
Catalog No. 23-246-390 Supplier Thermo Scientific™ Supplier No. R686010
Add to Cart
Add to Cart
Mitsubishi™ Double Zip AnaeroPouch-Bag is used to hold AnaeroPouch-Anaero, AnaeroPouch-MicroAero, and AnaeroPouch-CO2 gas generators.
Mitsubishi™ Double Zip AnaeroPouch-Bag is used to hold AnaeroPouch-Anaero, AnaeroPouch-MicroAero, and AnaeroPouch-CO2 gas generators.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type AnaeroPouch-Bag, Double Zip
Format Bag
Includes Double Zip Bag
Product Line Thermo Scientific Mitsubishi
Quantity 20/Pk.