Learn More
Electron Microscopy Sciences EMS Delicate Double-Curve Dissecting Scissors
SDP
Catalog No. 501931261
Encompass_Preferred
These scissors are finger-friendly with a ribbon handle. Extra sharp, with single serrated edge. The double curvature of these scissors makes dissection easier in hard to reach areas or where a clear view is not always an option. Made in Germany of high grade stainless steel. They measure 4 5/8" (117.4 mm).
OAL: 4-5/8
Catalog No. 50-193-1261 Supplier Electron Microscopy Sciences Supplier No. 7299803
These scissors are finger-friendly with a ribbon handle. Extra sharp, with single serrated edge. The double curvature of these scissors makes dissection easier in hard to reach areas or where a clear view is not always an option. Made in Germany of high grade stainless steel. They measure 4 5/8" (117.4 mm).
OAL: 4-5/8