Learn More
Abcam DoubleStain CL IHC Kit: G&Rt on human/mouse tissue (HRP/Green & AP/Red), 60ML
Catalog No. 502673160
Encompass_Preferred
Abcam's Doublestain IHC Kit (ab183282) is designed to be used with user supplied goat and rat primary antibodies to detect two distinct antigens on human or mouse tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-3160 Supplier Abcam Supplier No. AB18328260ML
Abcam's Doublestain IHC Kit (ab183282) is designed to be used with user supplied goat and rat primary antibodies to detect two distinct antigens on human or mouse tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement