Learn More
Abcam DoubleStain IHC Kit: G&M on human tissue (Green/HRP & AP/Red), 60ML
Catalog No. 502673166
Encompass_Preferred
Abcam's DoubleStain IHC kit (ab183272) is designed to use with user supplied goat and mouse antibodies to detect two distinct antigens on human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-3166 Supplier Abcam Supplier No. AB18327260ML
Abcam's DoubleStain IHC kit (ab183272) is designed to use with user supplied goat and mouse antibodies to detect two distinct antigens on human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement