Learn More
Abcam DoubleStain IHC kit: G&M on human tissue (BCIP & AEC), 60ML
Catalog No. 502673169
Encompass_Preferred
The DoubleStain IHC kit (ab183271) is designed to use with user supplied goat and mouse antibodies to detect two distinct antigens on human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-3169 Supplier Abcam Supplier No. AB18327160ML
The DoubleStain IHC kit (ab183271) is designed to use with user supplied goat and mouse antibodies to detect two distinct antigens on human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement