Learn More
Abcam DoubleStain IHC kit: G&M on human tissue (BCIP & AEC), 18ML
Catalog No. 502673170
Encompass_Preferred
The DoubleStain IHC kit (ab183271) is designed to use with user supplied goat and mouse antibodies to detect two distinct antigens on human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-3170 Supplier Abcam Supplier No. AB18327118ML
The DoubleStain IHC kit (ab183271) is designed to use with user supplied goat and mouse antibodies to detect two distinct antigens on human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement