Learn More
Abcam DoubleStain IHC Kit: G&M on human tissue (Green/HRP & AP/Red), 6ML
Catalog No. 502673165
Encompass_Preferred
Abcam's DoubleStain IHC kit (ab183272) is designed to use with user supplied goat and mouse antibodies to detect two distinct antigens on human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-3165 Supplier Abcam Supplier No. AB1832726ML
Abcam's DoubleStain IHC kit (ab183272) is designed to use with user supplied goat and mouse antibodies to detect two distinct antigens on human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement