Learn More
Abcam DoubleStain IHC Kit: G&Rt on human/mouse tissue (BCIP & AEC), 60ML
Catalog No. 502677849
Encompass_Preferred
The DoubleStain IHC kit (ab183281) is designed to be used with user supplied goat and rat primary antibodies to detect two distinct antigens on human or mouse tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-7849 Supplier Abcam Supplier No. AB18328160ML
The DoubleStain IHC kit (ab183281) is designed to be used with user supplied goat and rat primary antibodies to detect two distinct antigens on human or mouse tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement