Learn More
Abcam DoubleStain IHC Kit: M&R on human tissue (HRP/Green & AP/Red), 60ML
Catalog No. 502588556
Encompass_Preferred
Abcam's DoubleStain IHC Kit: M&R on human tissue (HRP/Green & AP/Red) (ab210061) is designed to detect two distinct antigens on human tissue or cell samples using a simplified protocol with user-supplied mouse and rabbit antibodies.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-8556 Supplier Abcam Supplier No. AB21006160ML
Abcam's DoubleStain IHC Kit: M&R on human tissue (HRP/Green & AP/Red) (ab210061) is designed to detect two distinct antigens on human tissue or cell samples using a simplified protocol with user-supplied mouse and rabbit antibodies.
The product is subject to the following: Abcam Restricted Use Statement