Learn More
Abcam DoubleStain IHC Kit: M&Rt on mouse tissue (BCIP & AEC), 60ML
Catalog No. 502673163
Encompass_Preferred
Abcam's DoubleStain IHC Kit (ab183278) is designed to use with user supplied mouse and rat primary antibody to detect two distinct antigens on mouse tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-3163 Supplier Abcam Supplier No. AB18327860ML
Add to Cart
Add to Cart
Request a Quote
Abcam's DoubleStain IHC Kit (ab183278) is designed to use with user supplied mouse and rat primary antibody to detect two distinct antigens on mouse tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement