Learn More
Abcam DoubleStain IHC Kit: M&Rt on mouse tissue (BCIP & AEC), 18ML
Catalog No. 502673164
Encompass_Preferred
Abcam's DoubleStain IHC Kit (ab183278) is designed to use with user supplied mouse and rat primary antibody to detect two distinct antigens on mouse tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-3164 Supplier Abcam Supplier No. AB18327818ML
Abcam's DoubleStain IHC Kit (ab183278) is designed to use with user supplied mouse and rat primary antibody to detect two distinct antigens on mouse tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement