Learn More
Abcam DoubleStain IHC Kit: R&Rt on human/mouse tissue (Green/HRP & AP/Red), 60ML
Catalog No. 502590566
Encompass_Preferred
Abcam's Doublestain IHC Kit (ab183285) is designed for use with user supplied rabbit and rat primary antibodies to detect two distinct antigens on human and mouse tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0566 Supplier Abcam Supplier No. AB18328560ML
Abcam's Doublestain IHC Kit (ab183285) is designed for use with user supplied rabbit and rat primary antibodies to detect two distinct antigens on human and mouse tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement